CellTiter-Blue® Cell Viability Assay
Supplier: Promega
Certificates
About this item
The CellTiter-Blue® Cell Viability Assay provides a homogeneous, fluorescent method for monitoring cell viability. The assay is based on the ability of living cells to convert a redox dye (resazurin) into a fluorescent end product (resorufin). Nonviable cells rapidly lose metabolic capacity and thus do not generate a fluorescent signal.
- Add-incubate-measure format reduces the number of handling steps
- Fast, flexible, time-saving
- Amenable to multiplexing with other assays
Assay procedure involves adding the single reagent directly to cells cultured in serum-supplemented medium. After an incubation step, data can be recorded using either a plate-reading fluorometer or spectrophotometer.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together