Cleanroom goggles, BioClean Clearview™, sterile, for single use
111-0186
: Nitritex
- Type:Anti-fog, anti-scratch, sterile
- Colour:Transparent
- Environmentally Preferable:
- Lenses:Clear PC
- Pk:60
Manufactured from light weight PVC, the ultra soft single use goggles provide outstanding comfort. The specially designed indirect ventilation system allows good ventilation while reducing the incidence of particle release.
- Toughened PC lens offering protection against flying particles
- Lightweight frame with internal lateral vents
- Fitted with four LLDPE indirect ventilation caps
- Non-linting, adjustable silicon head band
Suitable for use in cleanroom class ISO 4.
: EN 166:2001 Optical Class 1, PPE Cat II, Sterility validated to ISO 11135-1-2007
: Double bagged in low linting Tyek/PE pouches.