Restriction enzymes, NotI, 10 U/μl
Supplier: Promega
Certificates
About this item
These NotI restriction enzyme recognises GC˅ GGCC GCA and CG CCGG^ CG sites and cuts best at 37 °C in its own unique buffer.
- Capable of digesting DNA in 15 minutes or less
- Blue and white cloning assay provides a higher level of quality control for enzymes used in cloning applications
- Restriction enzymes are assayed to ensure optimal performance in genomic analysis applications
Storage buffer: 10 mM Tris-HCl (pH 7,4), 0,1% Triton® X-100, 500 mM KCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Delivery: Kit includes (1×1 ml) buffer D10X, 1×150 µl bovine serum albumin (acetylated), 1×0,25 ml MULTI-CORE™ 10X buffer.
Specifications
- Application:
- Source:
- Environmentally Preferable:
Specifications
Frequently Bought Together