Transfer membranes, PVDF, Pierce™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Transfer membranes made of high quality PVDF, which provides high binding capacity for proteins and nucleic acids.
- Ideal for colorimetric and chemiluminescent detection
- Resistent to discolouration and have better retention of absorbed protein
- Lower background levels and increased sensitivity for fluorescent probing
Compatible with most organic solvents, acids and mild bases; doesn't tear or become brittle like nitrocellulose.
Frequently Bought Together