Anti-KIRREL2 Antibody Pair
Catalog # ABNOH00084063-AP11
Supplier: Abnova
Specifications
- Antigen name:Kin of IRRE Like 2
- Antigen symbol:KIRREL2
- Host:Multiple
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:84063
- Antigen synonyms:FILTRIN|NEPH3|NLG1
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human KIRREL2.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-KIRREL2 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-KIRREL2, IgG2a kappa (20 µg);
Type:
Antigen: KIRREL2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human