Anti-Lymphoid Enhancer Factor 1 Mouse Monoclonal Antibody [clone: 6D538]
Catalog # USBIL8005-01-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Lymphoid Enhancer Factor 1
- Antigen symbol:LEF1
- Clonality:Monoclonal
- Clone:6D538
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Amino acid number:1 to 85
- Storage buffer:PBS, 0,1% sodium azide, PEG and sucrose, before the addiition of glycerol to 40%.
- Storage temperature:–20 °C
- Concentration:~0,1 mg/ml
- Immunogen:Recombinant LEF (Lymphoid Enhancer Binding Factor) (aa 1-85).
- Purification:purified by size exclusion chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 1 ?g/ml
Type: Primary
Antigen: Lymphoid Enhancer Factor 1
Clonality: Monoclonal
Clone: 6D538
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse,