Anti-Band 3 Protein Mouse monoclonal antibody unconjugated [clone: 8J283]
USBIB0072-01A
- Antibody type:Primary
- Antigen symbol:BAND3
- Clonality:Monoclonal
- Clone:8J283
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:5000
Type: Primary
Antigen: Band 3 Protein
Clonality: monoclonal
Clone: 8J283
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human