Anti-APIP Rabbit Polyclonal Antibody
NOVUNB100-56383
- Antibody type:Primary
- Antigen name:APAF1 interacting protein
- Antigen symbol:APIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:51074
- Antigen synonyms:MMRP19|CGI-29|CGI29|hAPIP|APIP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: APIP (APAF1 interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human