Order Entry
United States
ContactUsLinkComponent
Protein A/G magnetic beads
Pierce™ Protein A/G Magnetic Beads, Thermo Scientific
Catalog # PI88803
Supplier:  Invitrogen
undefined
Pierce™ Protein A/G Magnetic Beads, Thermo Scientific
Catalog # PI88803
Supplier:  Invitrogen
Supplier Number:  88803
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Protein A/G magnetic beads
  • Size:
    5 ml
  • Environmentally Preferable:
  • Cat. No.:
    PI88803
  • Supplier no.:
    88803

Specifications

About this item

Thermo Scientific Pierce Protein A/G Magnetic Beads are high-performance affinity particles for antibody purification and immunoprecipitation methods using manual or robotic magnetic separators.

  • High capacity-nearly four times higher binding capacity than typical magnetic beads from other suppliers, allowing the use of smaller amounts per experiment
  • Low non-specific binding-stable, pre-blocked beads provide clean purification products (e.g., antigen eluted in IP with antibody is devoid of contaminating proteins from complex IP matrix)
  • Flexibility-convenience of IgG binding domains of both Protein A and Protein G on one bead
  • Compatibility-beads are compatible with manual and automated applications (e.g., Thermo Scientific KingFisher Instruments)
  • Assay consistency-magnetic beads eliminate resin loss and provide for more efficient separation of solutions than traditional IP methods that use only microcentrifuge tubes

These magnetic beads are coated with genetically engineered Pierce Protein A/G, a recombinant fusion protein which combines the IgG binding domains of both Protein A and Protein G.

The recombinant Protein A/G that is immobilized onto the Pierce Magnetic Beads is a fusion of the IgG binding domains of both Protein A and Protein G. Protein A/G contains four Fc-binding domains from Protein A and two from Protein G, making it a convenient tool for investigating and purifying immunoglobulins. Thus, Pierce Magnetic Particles are not simply a mixed immobilization of separate Protein A and Protein G polypeptides, nor are they a mixture of Protein A magnetic beads and Protein G magnetic beads.

This enables capture of antibodies from a wider range of species and isotypes than either protein alone. Using our crosslinker chemistry, you can immobilize an antibody onto the magnetic particle and prevent IgG contamination in your immunoprecipitated sample. These beads can be used both manually with a magnetic stand as well as with automated platforms such as the Thermo Scientific KingFisher Instruments.

Ordering information: Upon receipt store at 4 °C. Product shipped with an ice pack.
Caution: For Research Use Only. Not for use in diagnostic procedures.