Anti-SMARCA4 Rabbit Polyclonal Antibody
Catalog # 77800-295
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Transcription activator BRG1
- Antigen Symbol:SMARCA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P51532
- Antigen Synonyms:ATP dependent helicase SMARCA4|ATP-dependent helicase SMARCA4|BAF 190|BAF190|BAF190A|Brahma protein like 1|BRG1|BRG1 associated factor 190A|BRG1 protein|BRG1-associated factor 190A|BRM/SWI2 related gene 1|Global transcription activator homologous sequence|global transcription activator snf2l4|Homeotic gene regulator|hSNF2b|Mitotic growth and transcription activator|MRD16|Nuclear protein GRB1|Protein brahma homolog 1|Protein BRG-1|Protein BRG1|RTPS2|SMARC A4|SMARCA4|SMCA4_HUMAN|SNF2|SNF2 beta|SNF2 like 4|SNF2-beta|SNF2B|SNF2L4|SNF2LB|Sucrose nonfermenting like 4|SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 4|SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4|SWI2|Transcription activator BRG1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SMARCA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat