Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: SLC22A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77820-370
- 77820-371
- Antigen Symbol:
- SLC22A3
- SLC22A3
- Antigen Name:
- Synthetic peptide of SLC22A3
- Synthetic peptide of SLC22A3
- Antigen Synonyms:
- EMT|EMT organic cation transporter 3|EMTH|Extraneuronal monoamine transporter|OCT 3|OCT3|Orct 3|Orct3|Organic cation transporter 3|S22A3_HUMAN|SLC22 A3|SLC22A 3|Slc22a3|Solute carrier family 22 (extraneuronal monoamine transporter) member 3|Solute carrier family 22 (organic cation transporter) member 3|Solute carrier family 22 member 3
- EMT|EMT organic cation transporter 3|EMTH|Extraneuronal monoamine transporter|OCT 3|OCT3|Orct 3|Orct3|Organic cation transporter 3|S22A3_HUMAN|SLC22 A3|SLC22A 3|Slc22a3|Solute carrier family 22 (extraneuronal monoamine transporter) member 3|Solute carrier family 22 (organic cation transporter) member 3|Solute carrier family 22 member 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75751
- O75751
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice