To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Sphingosine 1-phosphate receptor 1 protein (311-382AA)
- Antigen Symbol:S1PR1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P21453
- Antigen Synonyms:CD363| CHEDG 1| CHEDG1| D1S3362| ECGF 1| ECGF1| EDG 1| EDG1| endothelial differentiation G protein coupled receptor 1| Endothelial differentiation G-protein coupled receptor 1| Endothelial differentiation sphingolipid G protein coupled receptor 1| FLJ58121| G protein coupled sphingolipid receptor| g protein-coupled receptor edg-1| S1P receptor 1| S1P receptor Edg 1| S1P receptor Edg-1| S1P receptor Edg1| S1P(1) receptor| S1P1| s1pr1| S1PR1_HUMAN| sphingolipid g-protein-coupled receptor 1| Sphingosine 1 phosphate receptor Edg 1| Sphingosine 1 phosphate receptor EDG1| sphingosine 1- phosphate receptor 1| Sphingosine 1-phosphate receptor 1| Sphingosine 1-phosphate receptor Edg-1| Sphingosine 1phosphate receptor type 1 S1P1
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: S1PR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse