Order Entry
United States
ContactUsLinkComponent
Anti-RGS16 Rabbit Polyclonal Antibody
Anti-RGS16 Rabbit Polyclonal Antibody
Catalog # 77807-376
Supplier:  AFG Bioscience
New Product
Anti-RGS16 Rabbit Polyclonal Antibody
Catalog # 77807-376
Supplier:  AFG Bioscience
Supplier Number:  A40253-50UG
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antigen Name:
    Synthesized peptide derived from RGS16 around the non-phosphorylation site of Y168.
  • Antigen Symbol:
    RGS16
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse,
    Rat
  • Size:
    50 µg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    O15492
  • Antigen Synonyms:
    A28 RGS14|A28 RGS14P|A28-RGS14P|HGNC:9997|hRGS-r|OTTHUMP00000033147|Regulator of G protein signaling 16|Regulator of G protein signalling 16|Regulator of G-protein signaling 16|Retinal-specific RGS|Retinally abundant regulator of G protein signaling|Retinally abundant regulator of G-protein signaling|RGS 16|RGS R|RGS-R|Rgs14|RGS16|RGS16_HUMAN|RGSR
  • Storage Buffer:
    PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
  • Storage Temperature:
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Purification:
    Antigen affinity purified
  • Packaging:
    Vial

Specifications

About this item

Recommended dilution: IHC:1:100 to 1:300, IF:1:200 to 1:1000, ELISA:1:20000

Type:
Antigen: RGS16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat