Anti-KDM4C Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: KDM4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77834-543
- 77834-542
- Antigen Symbol:
- KDM4C
- KDM4C
- Antigen Name:
- Synthetic peptide of KDM4C
- Synthetic peptide of KDM4C
- Antigen Synonyms:
- bA146B14.1|GASC 1 protein|GASC-1 protein|GASC1|Gene amplified in squamous cell carcinoma 1 protein|JHDM3C|JmjC domain containing histone demethylation protein 3C|JmjC domain-containing histone demethylation protein 3C|JMJD2C|Jumonji domain containing 2C|Jumonji domain containing protein 2C|Jumonji domain-containing protein 2C|Kdm4c|KDM4C_HUMAN|Lysine (K) specific demethylase 4C|Lysine demethylase 4C|Lysine-specific demethylase 4C|TDRD14C|Tudor domain containing 14C
- bA146B14.1|GASC 1 protein|GASC-1 protein|GASC1|Gene amplified in squamous cell carcinoma 1 protein|JHDM3C|JmjC domain containing histone demethylation protein 3C|JmjC domain-containing histone demethylation protein 3C|JMJD2C|Jumonji domain containing 2C|Jumonji domain containing protein 2C|Jumonji domain-containing protein 2C|Kdm4c|KDM4C_HUMAN|Lysine (K) specific demethylase 4C|Lysine demethylase 4C|Lysine-specific demethylase 4C|TDRD14C|Tudor domain containing 14C
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H3R0
- Q9H3R0
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice