To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Keratin 15
- Antigen Symbol:KRT15
- Clonality:Monoclonal
- Clone:LHK15
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Bovine
- Size:500 μl
- Environmentally Preferable:
- Gene ID:3866
- Antigen Synonyms:K1CO|CK15|K15
- Storage Buffer:PBS, pH 7.4 containing 0.2% BSA and 0.09% sodium azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:A 17-mer synthetic peptide from the C-terminus of Keratin 15
- Tested Applications:ICC/IF
- Purification:Protein A affinity purified
- Cat. No.:89352-992
- Supplier no.:GTX72325
Specifications
About this item
Mouse monoclonal antibody [LHK15] to Cytokeratin 15
For IF: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: KRT15
Clonality: Monoclonal
Clone: LHK15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Bovine