To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Centromere Protein F, 350/400Kda
- Antigen Symbol:CENPF
- Clonality:Monoclonal
- Clone:14C10 1D8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:1063
- Antigen Synonyms:Centromere Protein F|AH Antigen|CENP F Kinetochore Protein|CENF|CENPF|Mitosin
- Amino Acid Number:1759 to 2093
- Storage Buffer:Phosphate-buffered saline, pH 7.2, containing no preservatives
- Molecular Weight:350 kDa
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Protein fragment expressed in E. coli corresponding to amino acids 1759-2093.
- Tested Applications:ICC/IF, IHC-P, IHC-Fr, IP, WB
- Purification:Protein G affinity purified.
- Cat. No.:89364-704
- Supplier no.:GTX70137
Mouse monoclonal [14C10 1D8] to CENPF
For IHC-Fr: Use at an assay dependent dilution. For IHC-P: Use at an assay dependent dilution. For IP: Use at an assay dependent dilution. For WB: Use at a dilution of 1:500. Predicted molecular weight: 350 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: CENPF
Clonality: Monoclonal
Clone: 14C10 1D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: