To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Luciferase
- Antigen Symbol:Luciferase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Firefly
- Western Blot:Yes
- Environmentally Preferable:
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M sodium chloride, pH 7.2, containing 0.01% sodium azide
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Luciferase [ Photinus pyralis (Firefly)]
- Tested Applications:ELISA, ICC/IF, IHC, WB
- Purification:IgG fraction
- Cat. No.:89360-842
- Supplier no.:GTX20498
Specifications
About this item
Luciferase is a luminescent enzyme, mostly known for its role as a reporter protein
Type: Primary
Antigen: Firefly Luciferase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Firefly