Anti-TAL1 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Implicated in the genesis of hemopoietic malignancies. It may play an important role in hemopoietic differentiation. Serves as a positive regulator of erythroid differentiation.
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: TAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Size:100 μl
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Molecular Weight:34 kDa
- Purification:Purified by antigen-affinity chromatography.
- Storage Buffer:1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Tested Applications:IHC-P
- Isotype:IgG
- Host:Rabbit
- Concentration:1 mg/ml
- Cat. No.:89358-498
- Reactivity:Human
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:TAL1
- Gene ID:6886
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human TAL1. The exact sequence is proprietary.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:TAL-1|Stem cell protein|SCL|TAL1|TCL5|T-cell acute lymphocytic leukemia protein 1|T-cell leukemia/lymphoma protein 5
- Antigen Name:T-cell acute lymphocytic leukemia 1
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



