To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Lymphocyte Antigen 6 Complex Locus C1
- Antigen Symbol:Ly-6C
- Clonality:Monoclonal
- Clone:HK1.4
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2c
- Reactivity:Mouse
- Size:0.25 mg
- Environmentally Preferable:
- Gene ID:17067
- Antigen Synonyms:Ly 6c|Ly6c protein|Lymphocyte antigen 6 complex locus C|Lymphocyte antigen 6C|Lymphocyte antigen Ly 6C|Lymphocyte antigen 6 complex
- Storage Buffer:PBS
- Storage Temperature:Store at 4C. Do not freeze.
- Immunogen:L3 cloned CTL cells
- Purification:Protein A or G purified
- Cat. No.:102161-506
- Supplier no.:NBP1-28047
Specifications
About this item
Rat Monoclonal Ly-6C Antibody (HK1.4) [FITC]. Tested Applications: Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, In vitro assay. Tested Reactivity: Mouse.
Type: Primary
Antigen: LY6C
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse