To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Wingless-type MMTV Integration Site Family, member 16
- Antigen Symbol:Wnt16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:0.05 ml
- Environmentally Preferable:
- Gene ID:51384
- Antigen Synonyms:protein Wnt-16|wingless-type MMTV integration site family|member 16
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic 16 amino acid peptide from internal region of human WNT16.
- Purification:Immunogen affinity purified
- Cat. No.:102173-898
- Supplier no.:NBP1-44215
The Wnt16 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Wnt16. This antibody reacts with human. The Wnt16 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Wnt16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse