Order Entry
United States
ContactUsLinkComponent
Anti-URI Rabbit Polyclonal Antibody
Anti-URI Rabbit Polyclonal Antibody
Catalog # 102214-474
Supplier:  Novus Biologicals
Anti-URI Rabbit Polyclonal Antibody
Catalog # 102214-474
Supplier:  Novus Biologicals
Supplier Number:  NBP1-79413
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    URI
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    8725
  • Antigen Synonyms:
    NNX3RPB5-mediating protein|FLJ10575|subunit 5-mediating protein|URIRNA polymerase II|chromosome 19 open reading frame 2|Protein NNX3|RMPunconventional prefoldin RPB5 interactor|RNA polymerase II subunit 5-mediating protein
  • Storage Buffer:
    PBS and 2% Sucrose
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide directed towards the middle region of human C19orf2. Peptide sequence NGEYVPRKSILKSRSRENSVCSDTSESSAAEFDDRRGVLRSISCEEATCS.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102214-474
  • Supplier no.:
    NBP1-79413

Specifications

About this item

The URI Antibody from Novus Biologicals is a rabbit polyclonal antibody to URI. This antibody reacts with human. The URI Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: URI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human