Order Entry
United States
ContactUsLinkComponent
Anti-UBR4 Rabbit Polyclonal Antibody
Anti-UBR4 Rabbit Polyclonal Antibody
Catalog # 102162-592
Supplier:  Novus Biologicals
Anti-UBR4 Rabbit Polyclonal Antibody
Catalog # 102162-592
Supplier:  Novus Biologicals
Supplier Number:  NBP1-28724
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Ubiquitin Protein Ligase E3 Component N-recognin 4
  • Antigen Symbol:
    UBR4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    23352
  • Antigen Synonyms:
    ubiquitin protein ligase E3 component n-recognin 4|KIAA1307zinc finger|EC 6.3.2.-|N-recognin-4|600 kDa retinoblastoma protein-associated factor|KIAA0462E3 ubiquitin-protein ligase UBR4|UBR1 type 1|ZUBR1retinoblastoma-associated factor 600-like protein|RBAF600FLJ41863|Zinc finger UBR1-type protein 1|Retinoblastoma-associated factor of 600 kDa|p600|EC 6.3.2
  • Storage Buffer:
    TBS and 0.1% BSA
  • Storage Temperature:
    Store at 4C. Do not freeze.
  • Concentration:
    0.2 mg/ml
  • Immunogen:
    The immunogen recognized by this antibody maps to a region between residue 5133 and 5183 of human retinoblastoma-associated factor 600 using the numbering given in entry NP_065816.1 (GeneID 23352).
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102162-592
  • Supplier no.:
    NBP1-28724

Specifications

About this item

The UBR4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UBR4. This antibody reacts with human. The UBR4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative).

Type: Primary
Antigen: UBR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human