To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Tripartite Motif-containing 3
- Antigen Symbol:TRIM3/BERP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:10612
- Antigen Synonyms:tripartite motif containing 3|tripartite motif protein TRIM3|tripartite motif-containing protein 3|BERPRING finger protein 97|Brain-expressed RING finger protein|RNF97FLJ16135|tripartite motif-containing 3|RING finger protein 22|RNF22brain expressed ring finger|HAC1
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human tripartite motif-containing 3 (brain expressed ring finger protein) using the numbering given in entry NP_006449.2 (GeneID 10612).
- Purification:Immunogen affinity purified
- Cat. No.:102110-900
- Supplier no.:NB100-60400
The TRIM3 / BERP Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRIM3 / BERP. This antibody reacts with human. The TRIM3 / BERP Antibody has been validated for the following applications: Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100 - 1:500
Type: Primary
Antigen: TRIM3/BERP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human