Order Entry
United States
ContactUsLinkComponent
 
Anti-TIGAR/C12orf5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  102179-102
Anti-TIGAR/C12orf5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  102179-102
 :  NBP1-49534H
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    TP53 Induced Glycolysis Regulatory Phosphatase
  • Antigen Symbol:
    TIGAR/C12orf5
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    57103
  • Antigen Synonyms:
    EC 3.1.3.46|chromosome 12 open reading frame 5|probable fructose-2,6-bisphosphatase TIGAR|transactivated by NS3TP2 protein|TIGARTP53-induced glycolysis and apoptosis regulator
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C in the dark.
  • Immunogen:
    A genomic peptide made to an internal region of the human TIGAR protein (within residues 50-200). [Swiss-Prot Q9NQ88]
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102179-102
  • Supplier no.:
    NBP1-49534H

 

 

The TIGAR / C12orf5 Antibody [HRP] from Novus Biologicals is a rabbit polyclonal antibody to TIGAR / C12orf5. This antibody reacts with human, mouse. The TIGAR / C12orf5 Antibody [HRP] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TIGAR/C12orf5
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse