To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:TIF1 alpha
- Antigen Symbol:TIF1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:8805
- Antigen Synonyms:EC 6.3.2.-|RNF82Tif1a|PTC6|TIF1ATIF1ALPHA|RING finger protein 82|E3 ubiquitin-protein ligase TRIM24|hTIF1|transcription intermediary factor 1-alpha|tripartite motif-containing 24|TIF1TF1A|Tripartite motif-containing protein 24|TIF1-alpha|transcriptional intermediary factor 1|EC 6.3.2|tripartite motif containing 24
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human Transcriptional Intermediary Factor 1 Alpha (Tripartite Motif-Containing 24 ) using the numbering given in entry NP_056989.2 (GeneID 8805).
- Tested Applications:, Proximity Ligation Assay
- Purification:Immunogen affinity purified
- Cat. No.:102107-874
- Supplier no.:NB100-2596
Specifications
About this item
The TIF1 alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to TIF1 alpha. This antibody reacts with human, mouse. The TIF1 alpha Antibody has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay.
Type: Primary
Antigen: TIF1 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse