To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Trans-Golgi Network Integral Membrane Protein 2
- Antigen Symbol:TGN46
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:10618
- Antigen Synonyms:Trans golgi network protein 2|Trans Golgi network protein TGN51|Trans Golgi network integral membrane protein 2|Trans-Golgi network integral membrane protein 2|TGN38|TGN 48|trans golgi network 46|TGN 51|TGN38 homolog|Trans-Golgi network protein 2|TGN-46|TGN51|TGN48|TGOLN2|TGN-38|TGN 46|TTGN 2|TGOLN 2|TTGN2|Trans Golgi network protein (46 48 51kD isoforms)|Trans Golgi network integral membrane protein 2 [Precursor]|trans golgi network 38|Trans-Golgi network protein TGN51|TGN46
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4 °C in the dark.
- Immunogen:A genomic peptide made to an internal region of the human TGN46 protein (within residues 200-350). [Swiss-Prot O43493]
- Purification:Immunogen affinity purified
- Cat. No.:102179-336
- Supplier no.:NBP1-49643C
Specifications
About this item
The TGN46 Antibody [DyLight 650] from Novus Biologicals is a rabbit polyclonal antibody to TGN46. This antibody reacts with human. The TGN46 Antibody [DyLight 650] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:400
Type: Primary
Antigen: TGN46
Clonality: Polyclonal
Clone:
Conjugation: DyLight 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human