Order Entry
United States
ContactUsLinkComponent
 
Anti-TFIIE-alpha Rabbit Polyclonal Antibody
Catalog # 102175-776
Supplier:  Novus Biologicals
Anti-TFIIE-alpha Rabbit Polyclonal Antibody
Catalog # 102175-776
Supplier:  Novus Biologicals
Supplier Number:  NBP1-46195
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    TFIIE-alpha
  • Antigen Symbol:
    TFIIE-alpha
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    2960
  • Antigen Synonyms:
    General transcription factor IIE 56 kDa subunit|alpha 56kDa|TFIIE-A|56kD)|TF2E1|general transcription factor IIE|polypeptide 1|Transcription initiation factor IIE subunit alpha|FE|polypeptide 1 (alpha subunit|TFIIE-alpha|general transcription factor IIE subunit 1
  • Storage Buffer:
    Tris-Citrate/Phosphate (pH 7.0 - 8.0)
  • Storage Temperature:
    Store at 4C. Do not freeze.
  • Concentration:
    1.0 mg/ml
  • Immunogen:
    The immunogen for this product maps to a region between residue 100 and 150 of human General Transcription Factor IIE, polypeptide 1, alpha, using the numbering given in entry NP_005504.2 (GeneID 2960).
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102175-776
  • Supplier no.:
    NBP1-46195

Specifications

About this item

The TFIIE-alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to TFIIE-alpha. This antibody reacts with human. The TFIIE-alpha Antibody has been validated for the following applications: Immunohistochemistry, Immunoprecipitation, Western Blot (Negative).

Type: Primary
Antigen: TFIIE-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human