Order Entry
United States
ContactUsLinkComponent
Anti-CCL17/TARC Rabbit Polyclonal Antibody
Anti-CCL17/TARC Rabbit Polyclonal Antibody
Catalog # 102169-710
Supplier:  Novus Biologicals
Anti-CCL17/TARC Rabbit Polyclonal Antibody
Catalog # 102169-710
Supplier:  Novus Biologicals
Supplier Number:  NBP1-40991
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    TARC
  • Antigen Symbol:
    CCL17/TARC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Gene ID:
    6361
  • Antigen Synonyms:
    member 17|ABCD-2|T cell-directed CC chemokine|CC chemokine TARC|chemokine (C-C motif) ligand 17|small inducible cytokine subfamily A (Cys-Cys)|Thymus and activation-regulated chemokine|MGC138273|TARCA-152E5.3|C-C motif chemokine 17|Small-inducible cytokine A17|SCYA17MGC138271
  • Storage Buffer:
    PBS (pH 7.2)
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Concentration:
    1.0 mg/ml
  • Immunogen:
    Recombinant human CCL17
  • Purification:
    IgG purified
  • Cat. No.:
    102169-710
  • Supplier no.:
    NBP1-40991

Specifications

About this item

The CCL17 / TARC Antibody from Novus Biologicals is a rabbit polyclonal antibody to CCL17 / TARC. This antibody reacts with human. The CCL17 / TARC Antibody has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Functional.

Type: Primary
Antigen: CCL17/TARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human