Order Entry
United States
ContactUsLinkComponent
Anti-SLC20A2 Rabbit Polyclonal Antibody
Anti-SLC20A2 Rabbit Polyclonal Antibody
Catalog # 102199-320
Supplier:  Novus Biologicals
Anti-SLC20A2 Rabbit Polyclonal Antibody
Catalog # 102199-320
Supplier:  Novus Biologicals
Supplier Number:  NBP1-69702
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Solute Carrier Family 20 Member A2
  • Antigen Symbol:
    SLC20A2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Pig,
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    6575
  • Antigen Synonyms:
    MLVAR|PiT-2|Gibbon ape leukemia virus receptor 2|Phosphate transporter 2|sodium-dependent phosphate transporter 2|amphotropic|Solute carrier family 20 member 2|member 2|GLVR2PIT-2|Glvr-2|pit2|receptor for|solute carrier family 20 (phosphate transporter)|hPit2|murine leukemia virus
  • Storage Buffer:
    PBS & 2% Sucrose.
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptides corresponding to SLC20A2(solute carrier family 20 (phosphate transporter), member 2) The peptide sequence was selected from the N terminal of SLC20A2. Peptide sequence DVNLYNETVETLMAGEVSAMVGSAVWQLIASFLRLPISGTHCIVGSTIGF.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102199-320
  • Supplier no.:
    NBP1-69702

Specifications

About this item

The SLC20A2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC20A2. This antibody reacts with human, porcine. The SLC20A2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SLC20A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Porcine