To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:RNA Binding Motif Protein 25
- Antigen Symbol:RBM25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:58517
- Antigen Synonyms:RNA-binding protein 25|RNA-binding region (RNP1|fSAP94|NET52|RNA-binding motif protein 25|MGC105088|Arg/Glu/Asp-rich protein of 120 kDa|Protein S164|Arg/Glu/Asp-rich protein|RED120RNA-binding region-containing protein 7|RNA binding motif protein 25|RNPC7|functional spliceosome-associated protein 94|MGC117168|120 kDa|U1 small nuclear ribonucleoprotein 1SNRP homolog|RRM) containing 7|Snu71|S164
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 475 and 525 of human RNA binding motif protein 25 (Arg/Glu/Asp-rich protein, 120 kDa) using the numbering given in entry AAF19255.1 (GeneID 58517).
- Purification:Immunogen affinity purified
- Cat. No.:102110-454
- Supplier no.:NB100-57506
The RBM25 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RBM25. This antibody reacts with human, mouse. The RBM25 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100 - 1:500
Type: Primary
Antigen: RBM25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse