To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin Interaction Motif Containing 1
- Antigen Symbol:RAP80
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:51720
- Antigen Synonyms:RAP80X2HRIP110|Ubiquitin interaction motif-containing protein 1|Receptor-associated protein 80|ubiquitin interaction motif containing 1|RXRIP110|receptor associated protein 80|Retinoid X receptor-interacting protein 110|retinoid x receptor interacting protein|BRCA1-A complex subunit RAP80
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human RAP80. The immunogen is located within amino acids 30 - 80 of RAP80.
- Purification:Immunogen affinity purified
- Cat. No.:102209-528
- Supplier no.:NBP1-76829
Specifications
About this item
The RAP80 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RAP80. This antibody reacts with human, mouse. The RAP80 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: RAP80
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse