To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Poly-U Binding Splicing Factor 60KDa
- Antigen Symbol:PUF60
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:22827
- Antigen Synonyms:poly-U binding splicing factor 60KDa|FUSE-binding protein-interacting repressor|Ro-binding protein 1|FLJ31379|Ro ribonucleoprotein binding protein 1|SIAHBP1pyrimidine tract binding splicing factor|siah-BP1|ROBPI|poly(U)-binding-splicing factor PUF60|roBP1|siah binding protein 1,60 kDa poly(U)-binding-splicing factor|Siah-binding protein 1|FIRpoly-U binding splicing factor PUF60|FBP-interacting repressor|Ro ribonucleoprotein-binding protein 1|FBP interacting repressor
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen for this product maps to a region between residue 509 and 559 of human Poly-U Binding Splicing Factor 60KDa using the numbering given in entry NP_510965.1 (GeneID 22827).
- Purification:Immunogen affinity purified
- Cat. No.:102179-738
- Supplier no.:NBP1-49906
The PUF60 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PUF60. This antibody reacts with human, mouse. The PUF60 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative).
Type: Primary
Antigen: PUF60
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse