To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Pre-mRNA Processing Factor 3
- Antigen Symbol:PRPF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:9129
- Antigen Synonyms:PRP3 pre-mRNA processing factor 3 homolog (S. cerevisiae)|U4/U6 snRNP 90 kDa protein|PRP3|Prp3p|PRP3 pre-mRNA processing factor 3 homolog (yeast)|RP18|Pre-mRNA-splicing factor 3|hPrp3|HPRP3retinitis pigmentosa 18 (autosomal dominant)|U4/U6 small nuclear ribonucleoprotein Prp3|U4/U6-associated RNA splicing factor|HPRP3P
- Storage Buffer:PBS & 2% Sucrose.
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to PRPF3 (PRP3 pre-mRNA processing factor 3 homolog (S. cerevisiae)) The peptide sequence was selected from the N terminal of PRPF3. Peptide sequence VDKLFEAVEEGRSSRHSKSSSDRSRKRELKEVFGDDSEISKESSGVKKRR.
- Purification:Immunogen affinity purified
- Cat. No.:102189-302
- Supplier no.:NBP1-57201
Specifications
About this item
The PRPF3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PRPF3. This antibody reacts with human. The PRPF3 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: PRPF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human