Order Entry
United States
ContactUsLinkComponent
Anti-OAZ2 Rabbit Polyclonal Antibody
Anti-OAZ2 Rabbit Polyclonal Antibody
  102199-924
Anti-OAZ2 Rabbit Polyclonal Antibody
  102199-924
 :  NBP1-70661
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Ornithine decarboxylase antizyme 2
  • Antigen Symbol:
    OAZ2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    4947
  • Antigen Synonyms:
    ODC-Az 2|ornithine decarboxylase antizyme 2|AZ2
  • Storage Buffer:
    PBS and 2% Sucrose
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptides corresponding to OAZ2(ornithine decarboxylase antizyme 2) The peptide sequence was selected from the middle region of OAZ2. Peptide sequence PDGLLADGSKEGLLALLEFAEEKMKVNYVFICFRKGREDRAPLLKTFSFL.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102199-924
  • Supplier no.:
    NBP1-70661

 

 

The OAZ2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to OAZ2. This antibody reacts with human. The OAZ2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: OAZ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human