Order Entry
United States
ContactUsLinkComponent
Anti-MNAT1/MAT2A Rabbit Polyclonal Antibody
Anti-MNAT1/MAT2A Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-MNAT1/MAT2A Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 102126-382
  • 102126-396
  • Antigen Symbol:
  • MAT1/2A
  • MAT1/2A
  • Antigen Name:
  • MNAT1, CDK activating kinase assembly factor and Methionine Adenosyltransferase 2A
  • MNAT1, CDK activating kinase assembly factor and Methionine Adenosyltransferase 2A
  • Antigen Synonyms:
  • Methionine adenosyltransferase II|alpha|MAT 2|MATA2SAMS2adoMet synthetase 2|Methionine adenosyltransferase 2|methionine adenosyltransferase II|MAT-II|AMS2|S-adenosylmethionine synthase isoform type-2|S-adenosylmethionine synthetase isoform type-2|MATII|EC 2.5.1.6|AdoMet synthase 2
  • Methionine adenosyltransferase II|alpha|MAT 2|MATA2SAMS2adoMet synthetase 2|Methionine adenosyltransferase 2|methionine adenosyltransferase II|MAT-II|AMS2|S-adenosylmethionine synthase isoform type-2|S-adenosylmethionine synthetase isoform type-2|MATII|EC 2.5.1.6|AdoMet synthase 2
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Zebrafish, Primates, Human, Bovine, Rat
  • Zebrafish, Primates, Human, Bovine, Rat
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 4144
  • 4144
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide made to an internal portion of the human MAT2A protein (within residues 100-200). [Swiss-Prot# P31153]
  • Synthetic peptide made to an internal portion of the human MAT2A protein (within residues 100-200). [Swiss-Prot# P31153]
  • Purification:
  • Immunogen affinity purified
  • Immunogen affinity purified
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Storage Buffer:
  • PBS and 30% Glycerol
  • PBS and 30% Glycerol
  • Storage Temperature:
  • Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.

Specifications

Related Information