To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:Hrk
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:8739
- Antigen Synonyms:Neuronal death protein DP5|harakiri|BCL2 interacting protein (contains only BH3 domain)|DP5|death protein 5|BCL2-interacting protein (contains only BH3 domain)|activator of apoptosis harakiri|BH3-interacting domain-containing protein 3|BCL2-interacting protein|HARAKIRI|BID3|activator of apoptosis Hrk
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Hrk. The immunogen is located within the last 50 amino acids of Hrk.
- Purification:Immunogen affinity purified
- Cat. No.:102207-924
- Supplier no.:NBP1-76414
Rabbit Polyclonal Hrk Antibody. Tested Applications: Western Blot, ELISA. Tested Reactivity: Human, Mouse.
Type: Primary
Antigen: Hrk
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human