To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Fused in Sarcoma
- Antigen Symbol:FUS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:2521
- Antigen Synonyms:fusion|HNRNPP2|Oncogene TLS|FUS1|amyotrophic lateral sclerosis 6|fus-like protein|16) malignant liposarcoma|fusion gene in myxoid liposarcoma|fused in sarcoma|ALS6|Translocated in liposarcoma protein|TLShnRNP-P2|75 kDa DNA-pairing protein|fusion (involved in t(12|16) in malignant liposarcoma)|RNA-binding protein FUS|heterogeneous nuclear ribonucleoprotein P2|Oncogene FUS|POMp75|derived from t(12
- Storage Buffer:PBS & 2% Sucrose.
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to FUS(fusion (involved in t(12;16) in malignant liposarcoma)) The peptide sequence was selected from the N terminal of FUS. Peptide sequence MASNDYTQQATQSYGAYPTQPGQGYSQQSSQPYGQQSYSGYSQSTDTSGY.
- Purification:Immunogen affinity purified
- Cat. No.:102189-512
- Supplier no.:NBP1-57309
The FUS Antibody from Novus Biologicals is a rabbit polyclonal antibody to FUS. This antibody reacts with human. The FUS Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human