Order Entry
United States
ContactUsLinkComponent
 
Anti-EIF2 beta Rabbit Polyclonal Antibody
  102120-706
Anti-EIF2 beta Rabbit Polyclonal Antibody
  102120-706
 :  NB100-93284
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Eukaryotic Initiation Factor 2 Beta
  • Antigen Symbol:
    EIF2 beta
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    8894
  • Antigen Synonyms:
    38kDa|EIF2|eukaryotic translation initiation factor 2|eukaryotic initiation factor 2-beta|subunit 2 beta|EIF2beta|EIF2B|Eukaryotic translation initiation factor 2 subunit beta|eukaryotic translation initiation factor 2 beta|subunit 2 (beta|38kD )|eIF-2-beta|MGC8508|eukaryotic translation initiation factor 2 subunit 2|DKFZp686L18198
  • Storage Buffer:
    TBS and 0.1% BSA
  • Storage Temperature:
    Store at 4C. Do not freeze.
  • Concentration:
    0.2 mg/ml
  • Immunogen:
    The immunogen recognized by this antibody maps to a region between residue 283 and 333 of human eukaryotic translation initiation factor 2, subunit 2 beta, 38kDa using the numbering given in entry NP_003899.2 (GeneID 8894).
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102120-706
  • Supplier no.:
    NB100-93284

 

 

The EIF2 beta Antibody from Novus Biologicals is a rabbit polyclonal antibody to EIF2 beta. This antibody reacts with human, mouse. The EIF2 beta Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: EIF2 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse