To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:Dicer
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:23405
- Antigen Synonyms:DCR1|Dicer|EC 3.1.26.3|HERNADicer1|Dcr-1 homolog (Drosophila)|KIAA0928endoribonuclease Dicer|double-stranded RNA-specific endoribonuclease|Helicase MOI|EC 3.1.26|dicer 1|ribonuclease type III|K12H4.8-LIKE|helicase-moi|Helicase with RNase motif|EC 3.1.26.-|Dcr-1 homolog|Dicer1
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 600 and 650 of human double-stranded RNA-specific endoribonuclease; dicer1 using the numbering given in entry NP_085124.2 (GeneID 23405).
- Purification:Immunogen affinity purified
- Cat. No.:102157-206
- Supplier no.:NBP1-06520
The Dicer Antibody from Novus Biologicals is a rabbit polyclonal antibody to Dicer. This antibody reacts with human, mouse. The Dicer Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
There is 1 scientific publication using this antibody.
Type: Primary
Antigen: Dicer
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse