To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:DEAD (Asp-Glu-Ala-Asp) box polypeptide 19B
- Antigen Symbol:DDX19B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:11269
- Antigen Synonyms:DEAD box protein 19B|DDX19DEAD-box protein 5|ATP-dependent RNA helicase DDX19|DEAD-box RNA helicase DEAD5|RNAh|homolog)|DEAD (Asp-Glu-Ala-As) box polypeptide 19|yeast Dbp5 homolog|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19 (Dbp5|EC 3.6.4.13|DEAD (Asp-Glu-Ala-As) box polypeptide 19B|yeast|DEAD box RNA helicase DEAD5|EC 3.6.1|DBP5ATP-dependent RNA helicase DDX19B
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 450 and the C-terminus (residue 479) of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 19B using the numbering given in entry NP_009173.1 (GeneID 11269).
- Purification:Immunogen affinity purified
- Cat. No.:102116-810
- Supplier no.:NB100-760
Specifications
About this item
The DDX19B Antibody from Novus Biologicals is a rabbit polyclonal antibody to DDX19B. This antibody reacts with human, mouse. The DDX19B Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: DDX19B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse