To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CaMKI gamma
- Antigen Symbol:CAMKIG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:57172
- Antigen Synonyms:VWS1CLICK III|CaM-KI gamma|caMKIG|CaM kinase I gamma|CaM kinase IG|CLICKIII|CaMK-like CREB kinase III|EC 2.7.11|dJ272L16.1|CaMKI gamma|calcium/calmodulin-dependent protein kinase IG|calcium/calmodulin-dependent protein kinase type 1G|EC 2.7.11.17
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to CAMK1G(calcium/calmodulin-dependent protein kinase IG) Antibody(against the N terminal of CAMK1G. Peptide sequence SEVFLVKQRLTGKLFALKCIKKSPAFRDSSLENEIAVLKKIKHENIVTLE.
- Purification:Immunogen affinity purified
- Cat. No.:102190-460
- Supplier no.:NBP1-57827
Specifications
About this item
The CaMKI gamma Antibody from Novus Biologicals is a rabbit polyclonal antibody to CaMKI gamma. This antibody reacts with human. The CaMKI gamma Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: CaMKI gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human