Order Entry
United States
ContactUsLinkComponent
 
Anti-ATOH8 Rabbit Polyclonal Antibody
  102211-562
Anti-ATOH8 Rabbit Polyclonal Antibody
  102211-562
 :  NBP1-77347
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Atonal homolog 8 (Drosophila)
  • Antigen Symbol:
    ATOH8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    84913
  • Antigen Synonyms:
    Helix-loop-helix protein hATH-6|BHLHA21|ATH6|basic helix-loop-helix transcription factor 6|FLJ38730|atonal homolog 8 (Drosophila)|Class A basic helix-loop-helix protein 21|atonal homolog 6|bHLHa21basic helix loop helix transcription factor 6|FLJ14708|protein atonal homolog 8|hATH6
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ATOH8. The immunogen is located within the last 50 amino acids of ATOH8.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102211-562
  • Supplier no.:
    NBP1-77347

 

 

The ATOH8 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ATOH8. This antibody reacts with human, mouse, rat. The ATOH8 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: ATOH8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat