Order Entry
United States
ContactUsLinkComponent
Anti-alpha Adaptin Rabbit Polyclonal Antibody
Anti-alpha Adaptin Rabbit Polyclonal Antibody
Catalog # 102198-234
Supplier:  Novus Biologicals
Anti-alpha Adaptin Rabbit Polyclonal Antibody
Catalog # 102198-234
Supplier:  Novus Biologicals
Supplier Number:  NBP1-68949
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    alpha Adaptin
  • Antigen Symbol:
    alpha Adaptin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    160
  • Antigen Synonyms:
    AP-2 complex subunit alpha-1|alpha 1|Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit|alpha A|CLAPA1adaptin|alpha 1 subunit|Clathrin assembly protein complex 2 alpha-A large chain|Adaptor protein complex AP-2 subunit alpha-1|large|adaptor-related protein complex 2|Alpha-adaptin A|Adapter-related protein complex 2 alpha-1 subunit|clathrin-associated/assembly/adaptor protein|ADTAAAP2-ALPHA|alpha1-adaptin|100 kDa coated vesicle protein A
  • Storage Buffer:
    PBS and 2% Sucrose
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptides corresponding to AP2A1 (adaptor-related protein complex 2, alpha 1 subunit) The peptide sequence was selected from the C terminal of AP2A1. Peptide sequence ILAKLKRKKGPGAGSALDDGRRDPSSNDINGGMEPTPSTVSTPSPSADLL.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102198-234
  • Supplier no.:
    NBP1-68949

Specifications

About this item

The alpha Adaptin Antibody from Novus Biologicals is a rabbit polyclonal antibody to alpha Adaptin. This antibody reacts with human. The alpha Adaptin Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: alpha Adaptin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human