To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Adrenocortical Dysplasia homolog
- Antigen Symbol:ACD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:65057
- Antigen Synonyms:Ptop|adrenocortical dysplasia homolog (mouse)|Pip1|TPP1|PIP1Tint1|PTOPTpp1|POT1 and TIN2-interacting protein|TINT1adrenocortical dysplasia protein homolog|TIN2 interacting protein 1|POT1 and TIN2 organizing protein
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to ACD(adrenocortical dysplasia homolog (mouse)) The peptide sequence was selected from the middle region of ACD (NP_001075955). Peptide sequence KNRPPFPRTGATRGAQEPCSVWEPPKRHRDGSAFQYEYEPPCTSLCARVQ.
- Purification:Immunogen affinity purified
- Cat. No.:102183-764
- Supplier no.:NBP1-53154
Specifications
About this item
The ACD Antibody from Novus Biologicals is a rabbit polyclonal antibody to ACD. This antibody reacts with human. The ACD Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ACD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human