To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:RB binding protein 4, chromatin remodeling factor
- Antigen Symbol:RBBP4/RbAp48
- Clonality:Monoclonal
- Clone:13D10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:5928
- Antigen Synonyms:Chromatin assembly factor I p48 subunit|Nucleosome-remodeling factor subunit RBAP48|MSI1 protein homolog|CAF-I p48|Chromatin assembly factor 1 subunit C|chromatin assembly factor/CAF-1 p48 subunit|histone-binding protein RBBP4|RBBP-4|Retinoblastoma-binding protein 4CAF-I 48 kDa subunit|NURF55|CAF-1 subunit C|RbAp48|Retinoblastoma-binding protein p48|retinoblastoma binding protein 4
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:The complete coding region (amino acids 1-425) of RbAp48 expressed in E. coli.
- Purification:Immunogen affinity purified
- Cat. No.:102170-010
- Supplier no.:NBP1-41201
Specifications
About this item
The RBBP4 / RbAp48 Antibody (13D10) from Novus Biologicals is a mouse monoclonal antibody to RBBP4 / RbAp48. This antibody reacts with human. The RBBP4 / RbAp48 Antibody (13D10) has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.
Type: Primary
Antigen: RBBP4/RbAp48
Clonality: Monoclonal
Clone: 13D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human