To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Meiotic Recombination 11
- Antigen Symbol:Mre11
- Clonality:Monoclonal
- Clone:12D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:4361
- Antigen Synonyms:HNGS1|MRE11 homolog A|ATLDmeiotic recombination (S. cerevisiae) 11 homolog A|double-strand break repair protein MRE11A|Meiotic recombination 11 homolog 1|Meiotic recombination 11 homolog A|DNA recombination and repair protein|MRE11 meiotic recombination 11 homolog A (S. cerevisiae)|AT-like disease|MRE11MRE11B|MRE11 homolog 1|endo/exonuclease Mre11
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.89 mg/ml
- Immunogen:Amino acids 182-582 of Mre11 expressed in E. coli.
- Purification:Protein G purified
- Cat. No.:102109-260
- Supplier no.:NB100-473
Specifications
About this item
Mouse Monoclonal Mre11 Antibody (12D7). Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: MRE11
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human