To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:KBTBD7
- Clonality:Monoclonal
- Clone:1G3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Primates,Human,Dog,Rat
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:84078
- Antigen Synonyms:DKFZP434E2318|kelch repeat and BTB (POZ) domain containing 7|kelch repeat and BTB domain-containing protein 7
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human KBTBD7 (NP_115514) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102232-220
- Supplier no.:NBP2-02742
Mouse Monoclonal KBTBD7 Antibody (1G3). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Rat, Canine, Primate.
Type: Primary
Antigen: KBTBD7
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human