To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:HAND1
- Clonality:Monoclonal
- Clone:1G10
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:9421
- Antigen Synonyms:autonomic nervous system and neural crestderivatives-expressed protein 1|Extraembryonic tissues|bHLHa27heart- and neural crest derivatives-expressed protein 1|heart|heart and neural crest derivatives expressed 1|Hxt|EHAND|Class A basic helix-loop-helix protein 27|Thing1|BHLHA27
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human HAND1 (NP_004812) produced in E.coli.
- Purification:Immunogen affinity purified
- Cat. No.:102227-892
- Supplier no.:NBP2-00576
Specifications
About this item
Mouse Monoclonal HAND1 Antibody (1G10). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human.
Type: Primary
Antigen: HAND1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human