To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ficolin-3
- Antigen Symbol:FCN3
- Clonality:Monoclonal
- Clone:FCN
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:8547
- Antigen Synonyms:ficolin-3|Hakata antigen|FCNHficolin 3|ficolin (collagen/fibrinogen domain-containing) 3 (Hakata antigen)|HAKA1MGC22543|Collagen/fibrinogen domain-containing protein 3|Collagen/fibrinogen domain-containing lectin 3 p35|H-ficolin|ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen)
- Storage Buffer:10mM Phosphate (pH 7.4) and 0.5M NaCl
- Storage Temperature:Store at 4C in the dark.
- Concentration:1.0 mg/ml
- Immunogen:H-Ficolin (Hakata antigen, Ficolin-3) is an innate immunity pattern recognition molecule found in human serum. H-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP) formed by carbohydrates (GlcNAc, GalNAc and fucose) activating complement through the lectin pathway by means of the associated MASPs ((MBL)-associated serine proteases). H-Ficolin consists of 35 kDa protein chains associated in triplet subunits which are again associated in high molecular weight oligomers by means of disulfide bonds at the N-terminal region.
- Purification:Protein A purified
- Cat. No.:102170-068
- Supplier no.:NBP1-41283
Specifications
About this item
The Ficolin-3 Antibody (FCN) from Novus Biologicals is a mouse monoclonal antibody to Ficolin-3. This antibody reacts with human. The Ficolin-3 Antibody (FCN) has been validated for the following applications: ELISA.
Type: Primary
Antigen: FCN3
Clonality: Monoclonal
Clone: FCN
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG Kappa
Reactivity: Human