Order Entry
United States
ContactUsLinkComponent
Anti-ENPP-3/CD203c Mouse Monoclonal Antibody [clone: NP4D6]
Anti-ENPP-3/CD203c Mouse Monoclonal Antibody [clone: NP4D6]
Catalog # 102174-178
Supplier:  Novus Biologicals
Anti-ENPP-3/CD203c Mouse Monoclonal Antibody [clone: NP4D6]
Catalog # 102174-178
Supplier:  Novus Biologicals
Supplier Number:  NBP1-44643
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Ectonucleotide pyrophosphatase/phosphodiesterase 3
  • Antigen Symbol:
    ENPP-3/CD203c
  • Clonality:
    Monoclonal
  • Clone:
    NP4D6
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    5169
  • Antigen Synonyms:
    Phosphodiesterase I/nucleotide pyrophosphatase 3|Phosphodiesterase I beta|dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3)|NPP3|PD-IBETA|phosphodiesterase-I beta|ectonucleotide pyrophosphatase/phosphodiesterase 3|E-NPP 3|ectonucleotide pyrophosphatase/phosphodiesterase family member 3|CD203c|CD203c antigen|gp130RB13-6|dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3)|PDNP3B10
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Store at 4C. Do not freeze.
  • Concentration:
    1 mg/ml
  • Immunogen:
    HEK-293 cells transfected with human CD203c
  • Purification:
    Protein A purified
  • Cat. No.:
    102174-178
  • Supplier no.:
    NBP1-44643

Specifications

About this item

The ENPP-3 / CD203c Antibody (NP4D6) from Novus Biologicals is a mouse monoclonal antibody to ENPP-3 / CD203c. This antibody reacts with human. The ENPP-3 / CD203c Antibody (NP4D6) has been validated for the following applications: Flow Cytometry, Immunocytochemistry / Immunofluorescence, CyTOF-ready.

Type: Primary
Antigen: ENPP-3/CD203c
Clonality: Monoclonal
Clone: NP4D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human